Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dichlorophenyl)maleamic acid, 97%

$40.65 - $507.96

Chemical Identifiers

Chemical Name or Material N-(2,4-Dichlorophenyl)maleamic acid
CAS 95695-47-9
Molecular Formula C10H7Cl2NO2
Formula Weight 260.07
MDL Number MFCD00082905
Synonym 4-(2,4-Dichloroanilino)-4-oxobut-2-enoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2494103

 
alfa aesar™
B2494103
1g Each for $40.65

AAB2494106

 
alfa aesar™
B2494106
5g Each for $138.85

AAB2494114

 
alfa aesar™
B2494114
25g Each for $507.96
Description & Specifications

Specifications

Formula Weight 260.07
Chemical Name or Material N-(2,4-Dichlorophenyl)maleamic acid
Assay 97%
Molecular Formula C10H7Cl2NO2
Melting Point 177° to 180°C
MDL Number MFCD00082905
CAS 95695-47-9
Quantity 1g
Synonym 4-(2,4-Dichloroanilino)-4-oxobut-2-enoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.