missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4-Dichlorobenzyl)methylamine, 95%

$44.09 - $192.77

Chemical Identifiers

CAS 5013-77-4
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.067
MDL Number MFCD00045185
InChI Key GUJXWKXDISDARD-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl methylamine, n-methyl-2,4-dichlorobenzylamine, benzenemethanamine, 2,4-dichloro-n-methyl, 2,4-dichlorophenyl methyl methyl amine, 2,4-dichloro-n-methylbenzylamine, 1-2,4-dichlorophenyl-n-methylmethanamine, 2,4-dichlorophenyl-n-methylmethanamine, n-2,4-dichlorobenzyl-n-methylamine, 1-2,4-dichlorophenyl-n-methyl-methanamine, 2,4-dichlorobenzyl methyl amine
PubChem CID 485424
IUPAC Name 1-(2,4-dichlorophenyl)-N-methylmethanamine
SMILES CNCC1=C(C=C(C=C1)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6340903 View Documents Alfa Aesar™
H6340903
1g Each for $44.09
 
AAH6340906 View Documents Alfa Aesar™
H6340906
5g Each for $192.77
 
Specifications

Chemical Identifiers

5013-77-4
190.067
GUJXWKXDISDARD-UHFFFAOYSA-N
485424
CNCC1=C(C=C(C=C1)Cl)Cl
C8H9Cl2N
MFCD00045185
2,4-dichlorobenzyl methylamine, n-methyl-2,4-dichlorobenzylamine, benzenemethanamine, 2,4-dichloro-n-methyl, 2,4-dichlorophenyl methyl methyl amine, 2,4-dichloro-n-methylbenzylamine, 1-2,4-dichlorophenyl-n-methylmethanamine, 2,4-dichlorophenyl-n-methylmethanamine, n-2,4-dichlorobenzyl-n-methylamine, 1-2,4-dichlorophenyl-n-methyl-methanamine, 2,4-dichlorobenzyl methyl amine
1-(2,4-dichlorophenyl)-N-methylmethanamine

Specifications

Colorless
5013-77-4
MFCD00045185
2,4-dichlorobenzyl methylamine, n-methyl-2,4-dichlorobenzylamine, benzenemethanamine, 2,4-dichloro-n-methyl, 2,4-dichlorophenyl methyl methyl amine, 2,4-dichloro-n-methylbenzylamine, 1-2,4-dichlorophenyl-n-methylmethanamine, 2,4-dichlorophenyl-n-methylmethanamine, n-2,4-dichlorobenzyl-n-methylamine, 1-2,4-dichlorophenyl-n-methyl-methanamine, 2,4-dichlorobenzyl methyl amine
CNCC1=C(C=C(C=C1)Cl)Cl
190.067
190.07
95%
1g
C8H9Cl2N
UN3259
GUJXWKXDISDARD-UHFFFAOYSA-N
1-(2,4-dichlorophenyl)-N-methylmethanamine
485424
Low Melting Solid
N-(2,4-Dichlorobenzyl)methylamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.