Learn More
N-(2,4-Dichlorobenzyl)methylamine, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6340903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-(2,4-Dichlorobenzyl)methylamine | |
Colorless | |
MFCD00045185 | |
UN3259 | |
GUJXWKXDISDARD-UHFFFAOYSA-N | |
1-(2,4-dichlorophenyl)-N-methylmethanamine | |
485424 | |
95% |
5013-77-4 | |
C8H9Cl2N | |
1 g | |
2,4-dichlorobenzyl methylamine, n-methyl-2,4-dichlorobenzylamine, benzenemethanamine, 2,4-dichloro-n-methyl, 2,4-dichlorophenyl methyl methyl amine, 2,4-dichloro-n-methylbenzylamine, 1-2,4-dichlorophenyl-n-methylmethanamine, 2,4-dichlorophenyl-n-methylmethanamine, n-2,4-dichlorobenzyl-n-methylamine, 1-2,4-dichlorophenyl-n-methyl-methanamine, 2,4-dichlorobenzyl methyl amine | |
CNCC1=C(C=C(C=C1)Cl)Cl | |
190.067 | |
190.07 | |
Low Melting Solid |
Chemical Identifiers
5013-77-4 | |
190.067 | |
GUJXWKXDISDARD-UHFFFAOYSA-N | |
485424 | |
CNCC1=C(C=C(C=C1)Cl)Cl |
C8H9Cl2N | |
MFCD00045185 | |
2,4-dichlorobenzyl methylamine, n-methyl-2,4-dichlorobenzylamine, benzenemethanamine, 2,4-dichloro-n-methyl, 2,4-dichlorophenyl methyl methyl amine, 2,4-dichloro-n-methylbenzylamine, 1-2,4-dichlorophenyl-n-methylmethanamine, 2,4-dichlorophenyl-n-methylmethanamine, n-2,4-dichlorobenzyl-n-methylamine, 1-2,4-dichlorophenyl-n-methyl-methanamine, 2,4-dichlorobenzyl methyl amine | |
1-(2,4-dichlorophenyl)-N-methylmethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures