Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4,6-Trimethylphenyl)thiourea, 99%

Chemical Identifiers

Chemical Name or Material N- (2,4,6-Trimethylphenyl)thiourea
CAS 91059-97-1
Molecular Formula C10H14N2S
Formula Weight 194.3
MDL Number MFCD00051488
UN Number UN2811
Beilstein 2642667
Synonym N-Mesitylthiourea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1208306

 
alfa aesar™
A1208306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1208314

 
alfa aesar™
A1208314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1208322

 
alfa aesar™
A1208322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.3
Chemical Name or Material N- (2,4,6-Trimethylphenyl)thiourea
Assay 99%
Molecular Formula C10H14N2S
MDL Number MFCD00051488
CAS 91059-97-1
UN Number UN2811
Beilstein 2642667
Synonym N-Mesitylthiourea
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 230° to 231°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.