Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,4,6-Trichlorophenyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-(2,4,6-Trichlorophenyl)thiourea
CAS 31118-87-3
Molecular Formula C7H5Cl3N2S
Formula Weight 255.55
MDL Number MFCD00041153
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2745803

 
alfa aesar™
H2745803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2745806

 
alfa aesar™
H2745806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 255.55
Chemical Name or Material N-(2,4,6-Trichlorophenyl)thiourea
Assay 97%
Molecular Formula C7H5Cl3N2S
MDL Number MFCD00041153
CAS 31118-87-3
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point ≈220°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.