Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,3-Dimethylphenyl)thiourea, 98%

Chemical Identifiers

Chemical Name or Material N-(2,3-Dimethylphenyl)thiourea
CAS 55752-58-4
Molecular Formula C9H12N2S
Formula Weight 180.27
MDL Number MFCD00041163
UN Number UN2811
Beilstein 3249041
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1132303

  SDS
alfa aesar™
L1132303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1132306

  SDS
alfa aesar™
L1132306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.27
Chemical Name or Material N-(2,3-Dimethylphenyl)thiourea
Assay 98%
Molecular Formula C9H12N2S
MDL Number MFCD00041163
CAS 55752-58-4
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 202° to 204°C
Beilstein 3249041
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280f-P301+P310-P330-P501a
Wear protective clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.