missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,3-Dimethylphenyl)-3-methylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H5639503

 View more versions of this product

Catalog No. AAH5639503



Description

Description

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

N-(2,3-Dimethylphenyl)-3-methylbenzamide
292870-37-2
MFCD00443894
MTCYJXIELVPXBW-UHFFFAOYSA-N
N-(2,3-dimethylphenyl)-3-methylbenzamide
966014
97%
1g
C16H17NO
n-2,3-dimethylphenyl-3-methylbenzamide, aronis25672, n-2,3-dimethylphenyl 3-methylphenyl carboxamide, n-2,3-dimethylphenyl-3-methylbenzamide
CC1=C(C(=CC=C1)NC(=O)C2=CC(=CC=C2)C)C
239.318
239.31

Chemical Identifiers

292870-37-2
239.318
MTCYJXIELVPXBW-UHFFFAOYSA-N
966014
CC1=C(C(=CC=C1)NC(=O)C2=CC(=CC=C2)C)C
C16H17NO
MFCD00443894
n-2,3-dimethylphenyl-3-methylbenzamide, aronis25672, n-2,3-dimethylphenyl 3-methylphenyl carboxamide, n-2,3-dimethylphenyl-3-methylbenzamide
N-(2,3-dimethylphenyl)-3-methylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents