Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[2-(2-Pyridyl)ethy]thiourea, 98+%


Quantity 1g

Manufacturer: alfa aesar™  L10843-03

 View more versions of this product

Catalog No. AAL1084303

  • / Each
 

Description & Specifications

Chemical Identifiers

Melting Point 92° to 96°C
Quantity 1g
Chemical Name or Material N-[2-(2-Pyridyl)ethy]thiourea
CAS 180403-26-3
Assay ≥98%
Molecular Formula C8H11N3S
Formula Weight 181.26g/mol
MDL Number MFCD00041228
Beilstein 7635310
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P280f
Wear protective clothing.

Specifications

Chemical Name or Material N-[2-(2-Pyridyl)ethy]thiourea
CAS 180403-26-3
Molecular Formula C8H11N3S
Formula Weight 181.26g/mol
MDL Number MFCD00041228
Beilstein 7635310
Accessories

Warning

GHS P Statement
P280f
Wear protective clothing.

GHS H Statement
H302
Harmful if swallowed.