Please login to your online account to display your discounted pricing

N-[(2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-yl)methyl]-N-methylamine, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material N-[(2, 2-dimethyl-2, 3-dihydro-1-benzofuran-7-yl)methyl]-N-methylamine
CAS 868755-46-8
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD08435850
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC27246CB

  SDS
fisher scientific company
CC27246CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC27246DA

  SDS
fisher scientific company
CC27246DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.27
Quantity 250mg
Assay 95%
CAS Max % 100.0
Packaging Amber Glass Bottle
Assay Percent Range 95%
Chemical Name or Material N-[(2, 2-dimethyl-2, 3-dihydro-1-benzofuran-7-yl)methyl]-N-methylamine
MDL Number MFCD08435850
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 94.5
CAS 868755-46-8
Molecular Formula C12H17NO
Safety and Handling

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.