Please login to your online account to display your discounted pricing

N-(2,2-Diethoxyethyl)-N-(2-thienylmethyl)amine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material N-(2, 2-diethoxyethyl)-N-(2-thienylmethyl)amine
CAS 113825-05-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H19NO2S
Formula Weight 229.34
MDL Number MFCD00173734
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S09640CB

  SDS
fisher scientific company
S09640CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S09640DA

  SDS
fisher scientific company
S09640DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.34
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material N-(2, 2-diethoxyethyl)-N-(2-thienylmethyl)amine
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C11H19NO2S
MDL Number MFCD00173734
CAS 113825-05-1
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.