Please login to your online account to display your discounted pricing

N-(2,2-Diethoxyethyl)-N-(2-thienylmethyl)amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  S09640DA

 View more versions of this product

Catalog No. S09640DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 229.34
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material N-(2, 2-diethoxyethyl)-N-(2-thienylmethyl)amine
Packaging Amber Glass Bottle
Assay Percent Range 97%
MDL Number MFCD00173734
CAS 113825-05-1
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C11H19NO2S
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material N-(2, 2-diethoxyethyl)-N-(2-thienylmethyl)amine
CAS 113825-05-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H19NO2S
Formula Weight 229.34
MDL Number MFCD00173734
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.