missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide, 98+%

Manufacturer:  Alfa Aesar™ H5461603

 View more versions of this product

Catalog No. AAH5461603



Description

Description

Applications
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide was used in the synthesis of methyl [phenyl 5-(2,2,2-trichloroethoxycarbonylamino)-3,5-dideoxy-2-thio-β-D-glycero-D-galacto-2-nonulopyranosid]-onate; and phenyl 3,4-di-O-benzyl-6-O-(tert-butyl-dimethyl-silyl)-1-selanyl-2-(2,2,2-trichloroethoxycarbonylamine)-β-D-galactopyranoside.

Solubility
Soluble in dioxane, THF, DMF, CH2Cl2, alcohols, and most common organic solvents.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications

Specifications

N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide
111°C to 113°C
66065-85-8
MFCD00075216
Soluble in dioxane,THF,DMF,CH2Cl2,alcohols,and most common organic solvents.
C1CC(=O)N(C1=O)OC(=O)OCC(Cl)(Cl)Cl
290.477
290.49
Moisture sensitive
1g
C7H6Cl3NO5
2,5-dioxopyrrolidin-1-yl 2,2,2-trichloroethyl carbonate
WBZXNGAFYBGQFE-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 2,2,2-trichloroethyl carbonate
4192178
≥98%

Chemical Identifiers

66065-85-8
290.477
WBZXNGAFYBGQFE-UHFFFAOYSA-N
4192178
C1CC(=O)N(C1=O)OC(=O)OCC(Cl)(Cl)Cl
C7H6Cl3NO5
MFCD00075216
2,5-dioxopyrrolidin-1-yl 2,2,2-trichloroethyl carbonate
(2,5-dioxopyrrolidin-1-yl) 2,2,2-trichloroethyl carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c

H302+H312+H332-H319-H335

TSCA : No

Recommended Storage : Keep cold

Documents