Please login to your online account to display your discounted pricing

Alfa Aesar™ N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine, 98%

$36.64 - $443.31

Chemical Identifiers

Chemical Name or Material N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
CAS 82717-96-2
Molecular Formula C15H21NO4
Formula Weight 279.35
MDL Number MFCD00209976
Beilstein 3619501
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1935403

 
alfa aesar™
L1935403
1g Each for $36.64

AAL1935406

 
alfa aesar™
L1935406
5g Each for $114.68

AAL1935414

 
alfa aesar™
L1935414
25g Each for $443.31
Description & Specifications

Specifications

Formula Weight 279.35
Chemical Name or Material N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine
Assay 98%
Molecular Formula C15H21NO4
MDL Number MFCD00209976
CAS 82717-96-2
Beilstein 3619501
Melting Point 150° to 152°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.