Please login to your online account to display your discounted pricing

N-1-Naphthylethylenediamine Dihydrochloride, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$261.40 - $746.67

Chemical Identifiers

Chemical Name or Material N-1-Naphthylethylenediamine Dihydrochloride
CAS 1465-25-4
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C10H7NHCH2CH2NH2·2HCl
Formula Weight 259.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003505

 
avantor performance
R70105
Wide Mouth Amber Glass Bottle 100g Each for $746.67

02003506

 
avantor performance
R70103
Wide Mouth Amber Glass Bottle 25g Each for $261.40
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 259.18
Chemical Name or Material N-1-Naphthylethylenediamine Dihydrochloride
Molecular Formula C10H7NHCH2CH2NH2·2HCl
CAS 1465-25-4
Packaging Wide Mouth Amber Glass Bottle
Quantity 100g

    For Nitrogen Dioxide Determination (ASTM D-1607)