Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(1-Naphthyl)thiourea, 97%

Chemical Identifiers

Chemical Name or Material N-(1-Naphthyl)thiourea
CAS 86-88-4
Molecular Formula C11H10N2S
Formula Weight 202.29
MDL Number MFCD00041824
UN Number UN1651
Beilstein 778118
Merck Index 14,722
Synonym ANTU
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1902409

  SDS
alfa aesar™
A1902409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1902430

  SDS
alfa aesar™
A1902430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 202.29
Health Hazard 3 GHS P Statement
P280h-P301+P330+P331-P309-P310
Wear protective gloves/clothing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Synonym ANTU
Odor Odorless
Chemical Name or Material N-(1-Naphthyl)thiourea
TSCA TSCA
Melting Point 185° to 191°C
Quantity 10g
MDL Number MFCD00041824
Molecular Formula C11H10N2S
CAS 86-88-4
Beilstein 778118
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NAPHTHYLTHIOUREA
UN Number UN1651
RTECS Number YT9275000
Assay 97%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H351
Fatal if swallowed.
Suspected of causing cancer.
Merck Index 14,722
Safety and Handling

Danger

GHS P Statement
P280h-P301+P330+P331-P309-P310
Wear protective gloves/clothing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H300-H351
Fatal if swallowed.
Suspected of causing cancer.