Please login to your online account to display your discounted pricing

N-(1-Naphthyl)-N-(phenylmethylene)amine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AC33318DA

 View more versions of this product

Catalog No. AC33318DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 231.3
Quantity 1g
CAS Max % 100.0
Assay Percent Range 97%
MDL Number MFCD02235774
CAS Min % 96.5
Packaging Amber Glass Bottle
Chemical Name or Material N-(1-naphthyl)-N-(phenylmethylene)amine
CAS 890-51-7
Molecular Formula C17H13N

Specifications

Chemical Name or Material N-(1-naphthyl)-N-(phenylmethylene)amine
CAS 890-51-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C17H13N
Formula Weight 231.3
MDL Number MFCD02235774