Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(1-Naphthyl)-N-phenylmethacrylamide, 98%

$278.03 - $278.03

Chemical Identifiers

Chemical Name or Material N-(1-Naphthyl)-N-phenylmethacrylamide
CAS 141029-31-4
Molecular Formula C20H17NO
Formula Weight 287.36g/mol
MDL Number MFCD03701557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5104503

 
alfa aesar™
H51045-03
1g Each for $278.03
Description & Specifications

Specifications

Quantity 1g
Melting Point 112° to 118°C
Quantity 1g
Chemical Name or Material N-(1-Naphthyl)-N-phenylmethacrylamide
CAS 141029-31-4
Assay 98%
Molecular Formula C20H17NO
Formula Weight 287.36g/mol
MDL Number MFCD03701557
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.