Please login to your online account to display your discounted pricing

N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, ACS Grade, LabChem™

$149.63 - $149.63

Chemical Identifiers

Chemical Name or Material N-(1-Naphthyl)-Ethylenediamine Dihydrochloride
CAS 1465-25-4
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C10H7NHCH2CH2NH2·2HCl
Formula Weight 259.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC175408

 
lab chem
LC175408
Amber Glass 25g Each for $149.63
Description & Specifications

Specifications

Formula Weight 259.18
Chemical Name or Material N-(1-Naphthyl)-Ethylenediamine Dihydrochloride
Storage Information Room Temperature
Purity Grade ACS Grade
Sensitivity Passes Test
Solubility Soluble in water
Linear Formula C10H7NHCH2CH2NH2·2HCl
CAS Max % 100
Physical Form Powder
Water ≤5%
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECS Number 215-981-2
Packaging Amber Glass
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
CAS 1465-25-4
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Color Light tan
Quantity 25g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.