missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS

Used for colorimetric determination of sulfonamide in blood

$47.78 - $414.49

Chemical Identifiers

CAS 1465-25-4
Molecular Formula C12H16Cl2N2
Molecular Weight (g/mol) 259.174
MDL Number MFCD00012556
InChI Key MZNYWPRCVDMOJG-UHFFFAOYSA-N
Synonym 1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, ccris 425, marshall's reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride, n-1-naphthyl ethylenediamine dihydrochloride, n-1-naphthylethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, unii-h734599kjl
PubChem CID 15106
ChEBI CHEBI:53452
IUPAC Name N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6321406 View Documents Alfa Aesar™
J6321406
5g Each for $47.78
 
AAJ6321414 View Documents Alfa Aesar™
J6321414
25g Each for $158.14
 
AAJ6321422 View Documents Alfa Aesar™
J6321422
100g Each for $414.49
 
Description

Description

Applications
Used for colorimetric determination of sulfonamide in bloodN-(1-Naphthyl)ethylenediamine dihydrochloride acts as a coupling agent for the spectrophotometric determination of aromatic amines, aminophenols, phenylenediamines, dinitroanilines, chloroanilines, thiols and sulfonamides. It is also used in the quantitative analysis of nitrate and nitrite in water samples by colorimetry. It is involved in the preparation of (N-1-naphthyl-ethylenediamine)-dichloroplatinum(II) by reacting with potassium tetrachloroplatinate.

Solubility
Soluble in water or 95% ethanol.Soluble in hot water, alcohol, dilute hydrochloric acid and acetone.

Notes
Hygroscopic. Light sensitive. Incompatible with acids, acid chlorides, acid anhydrides, halogen acids and oxidizing agents.
Specifications

Chemical Identifiers

1465-25-4
259.174
MZNYWPRCVDMOJG-UHFFFAOYSA-N
15106
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
C12H16Cl2N2
MFCD00012556
1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, ccris 425, marshall's reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride, n-1-naphthyl ethylenediamine dihydrochloride, n-1-naphthylethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, unii-h734599kjl
CHEBI:53452
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl

Specifications

Hygroscopic; Light sensitive
191°C to 199°C
1465-25-4
MFCD00012556
14,6409
Soluble in water or 95% ethanol;Soluble in hot water,alcohol,dilute hydrochloric acid and acetone.
C1=CC=C2C(=C1)C=CC=C2NCCN.Cl.Cl
259.174
CHEBI:53452
Powder
N-(1-Naphthyl)ethylenediamine dihydrochloride
Cream to White or Gray
5g
C12H16Cl2N2
3707471
1,2-ethanediamine, n-1-naphthalenyl-, dihydrochloride, 2-1-naphthylamino ethylamine 2hcl, bratton-marshall reagent, ccris 425, marshall's reagent, n-1-naphthalenyl-1,2-ethanediamine dihydrochloride, n-1-naphthyl ethylenediamine dihydrochloride, n-1-naphthylethylenediamine dihydrochloride, n1-naphthalen-1-yl ethane-1,2-diamine dihydrochloride, unii-h734599kjl
MZNYWPRCVDMOJG-UHFFFAOYSA-N
N'-naphthalen-1-ylethane-1,2-diamine;dihydrochloride
15106
259.18
ACS
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 215-981-2

RTECSNumber : KV5330000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen