Please login to your online account to display your discounted pricing

N-(1-Naphthyl)-Ethylenediamine Dihydrochloride, 0.1%, for Nitrogen (Nitrite), LabChem™

$62.37 - $101.37

Chemical Identifiers

Chemical Name or Material N-(1-Naphthyl)-Ethylenediamine Dihydrochloride
CAS 7732-18-5
CAS Max % 99.9
CAS 1465-25-4
CAS Max % 0.1
Name Note 0.1%, for Nitrogen (Nitrite)
Linear Formula C10H7NHCH2CH2NH2·2HCl
Formula Weight 259.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC175501

 
lab chem
LC175501
500mL Each for $101.37

LC175507

 
lab chem
LC175507
125mL Each for $62.37
Description & Specifications

Specifications

Formula Weight 259.18
CAS Max % 99.9
Identification Passes Test
Linear Formula C10H7NHCH2CH2NH2·2HCl
Storage Information Refrigerate
Packaging Amber Poly
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Physical Form Liquid
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Hydrogen chloride
CAS 7732-18-5
Name Note 0.1%, for Nitrogen (Nitrite)
CAS Max % 0.1
Solubility Soluble in water
Chemical Name or Material N-(1-Naphthyl)-Ethylenediamine Dihydrochloride
Quantity 500mL
Color Colorless or light tan
CAS 1465-25-4
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.