Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(1-Naphthyl)-3-nitrobenzenesulfonamide, 97%

Chemical Identifiers

Chemical Name or Material N-(1-Naphthyl)-3-nitrobenzenesulfonamide
CAS 419559-79-8
Formula Weight 328.34
MDL Number MFCD01136840
Beilstein 2768935
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH58592MC

 
alfa aesar™
H58592MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH58592ME

 
alfa aesar™
H58592ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 328.34
Chemical Name or Material N-(1-Naphthyl)-3-nitrobenzenesulfonamide
Physical Form Solid
Assay 97%
MDL Number MFCD01136840
CAS 419559-79-8
Beilstein 2768935
Color Pale gray
Quantity 100mg