missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(1-Naphthyl)-2-nitrobenzenesulfonamide, 97%

$220.53 - $703.20

Chemical Identifiers

CAS 50393-59-4
Molecular Formula C16H12N2O4S
Molecular Weight (g/mol) 328.342
MDL Number MFCD00583392
InChI Key YPAFWESEDDOFFH-UHFFFAOYSA-N
Synonym n-1-naphthyl-2-nitrobenzenesulfonamide, n-naphthalen-1-yl-2-nitrobenzenesulfonamide, naphthyl 2-nitrophenyl sulfonyl amine, n-1-naphthyl-2-nitrobenzenesulfonamide, 2-nitro-n-1-naphthyl benzenesulfonamide
PubChem CID 681787
IUPAC Name N-naphthalen-1-yl-2-nitrobenzenesulfonamide
SMILES C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60668MD View Documents Alfa Aesar™
H60668MD
250mg Each for $220.53
 
AAH6066803 View Documents Alfa Aesar™
H6066803
1g Each for $703.20
 
Specifications

Chemical Identifiers

50393-59-4
328.342
YPAFWESEDDOFFH-UHFFFAOYSA-N
681787
C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3[N+](=O)[O-]
C16H12N2O4S
MFCD00583392
n-1-naphthyl-2-nitrobenzenesulfonamide, n-naphthalen-1-yl-2-nitrobenzenesulfonamide, naphthyl 2-nitrophenyl sulfonyl amine, n-1-naphthyl-2-nitrobenzenesulfonamide, 2-nitro-n-1-naphthyl benzenesulfonamide
N-naphthalen-1-yl-2-nitrobenzenesulfonamide

Specifications

250mg
C16H12N2O4S
3002217
YPAFWESEDDOFFH-UHFFFAOYSA-N
N-naphthalen-1-yl-2-nitrobenzenesulfonamide
681787
97%
50393-59-4
MFCD00583392
n-1-naphthyl-2-nitrobenzenesulfonamide, n-naphthalen-1-yl-2-nitrobenzenesulfonamide, naphthyl 2-nitrophenyl sulfonyl amine, n-1-naphthyl-2-nitrobenzenesulfonamide, 2-nitro-n-1-naphthyl benzenesulfonamide
C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3[N+](=O)[O-]
328.342
328.34
N-(1-Naphthyl)-2-nitrobenzenesulfonamide