missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(1-Glycyl-L-prolyl)alanine, 97%

$304.62 - $972.12

Chemical Identifiers

CAS 837-83-2
Molecular Formula C10H19N3O5
Molecular Weight (g/mol) 261.278
MDL Number MFCD00037344
InChI Key RLLPBIPOWCFAOP-LEUCUCNGSA-N
Synonym H-Gly-Pro-Ala-OH; Glycyl-L-prolyl-L-alanine
PubChem CID 91872902
IUPAC Name (2S)-2-[[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]amino]propanoic acid;hydrate
SMILES CC(C(=O)O)NC(=O)C1CCCN1C(=O)CN.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65592MD View Documents Alfa Aesar™
H65592MD
250mg Each for $304.62
 
AAH6559203 View Documents Alfa Aesar™
H6559203
1g Each for $972.12
 
Specifications

Chemical Identifiers

837-83-2
261.278
RLLPBIPOWCFAOP-LEUCUCNGSA-N
91872902
CC(C(=O)O)NC(=O)C1CCCN1C(=O)CN.O
C10H19N3O5
MFCD00037344
H-Gly-Pro-Ala-OH; Glycyl-L-prolyl-L-alanine
(2S)-2-[[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]amino]propanoic acid;hydrate

Specifications

837-83-2
MFCD00037344
H-Gly-Pro-Ala-OH; Glycyl-L-prolyl-L-alanine
CC(C(=O)O)NC(=O)C1CCCN1C(=O)CN.O
261.278
243.26
97%
250mg
C10H19N3O5
5288874
RLLPBIPOWCFAOP-LEUCUCNGSA-N
(2S)-2-[[(2S)-1-(2-aminoacetyl)pyrrolidine-2-carbonyl]amino]propanoic acid;hydrate
91872902
Powder
White
N-(1-Glycyl-L-prolyl)alanine
Safety and Handling

Safety and Handling

EINECSNumber : 212-654-6

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C