missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N,1-Di-Boc-L-histidine dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6257014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
160°C to 165°C (decomposition) | |
31687-58-8 | |
MFCD06804675 | |
WBGMQHNUPJENDC-MERQFXBCSA-N | |
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid | |
16211319 | |
98% | |
Solid |
N,1-Di-Boc-L-histidine dicyclohexylammonium salt | |
C28H48N4O6 | |
boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate | |
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2 | |
536.714 | |
536.7 | |
25 g |
Chemical Identifiers
31687-58-8 | |
536.714 | |
WBGMQHNUPJENDC-MERQFXBCSA-N | |
16211319 | |
CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(=O)OC(C)(C)C)C(=O)O.C1CCC(CC1)NC2CCCCC2 |
C28H48N4O6 | |
MFCD06804675 | |
boc-his boc-oh.dcha, boc-his boc-oh dcha, boc-his boc-ohdcha, boc-his boc .dcha, n,1-bis tert-butoxycarbonyl-l-histidine, compound with dicyclohexylamine 1:1, pubchem18934, n,1-bis boc-l-histidine dicyclohexylamine salt, boc-his boc-ohcha, boc-his boc-oh?cha, boc-his boc-oh toluene solvate | |
N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[1-[(2-methylpropan-2-yl)oxycarbonyl]imidazol-4-yl]propanoic acid |
Safety and Handling
EINECSNumber : 250-764-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only