missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride, ≥97%, Maybridge

Chemical Identifiers

CAS 849776-43-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H11NS·ClH
Formula Weight 213.73
MDL Number MFCD03306023
Chemical Name or Material N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC12296DA SDS Fisher Scientific Company
CC12296DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC12296DE Fisher Scientific Company
CC12296DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC12296CB SDS Fisher Scientific Company
CC12296CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

849776-43-8
100.0
C10H11NS·ClH
MFCD03306023
97%
96.5
97%
213.73
N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride

Specifications

213.73
100.0
MFCD03306023
Amber Glass Bottle
97%
97%
1g
C10H11NS·ClH
96.5
849776-43-8
N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.