Please login to your online account to display your discounted pricing

n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  CC01646CB

 View more versions of this product

Catalog No. CC01646CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 165.19
Quantity 250mg
Assay 97%
CAS 15205-27-3
Assay Percent Range 97%
Molecular Formula C9H11NO2
Chemical Name or Material N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine

Specifications

Chemical Name or Material N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H11NO2
Formula Weight 165.19