missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel, TCI America™

Manufacturer:  TCI America P17381G

 View more versions of this product

Catalog No. P17381G



Specifications

Specifications

[N-[1-[2-(2-Pyridylcarboxamido)phenyl]ethylidene]glycinato]nickel
1g
≥96.0% (T)
SNANRKUUXQKAKV-UHFFFAOYSA-L
nickel(2+);2-[1-[2-[[oxido(pyridin-2-yl)methylidene]amino]phenyl]ethylideneamino]acetate
53384302
Crystal-Powder at 20°C
Undesignated
264921-97-3
C16H13N3NiO3
CC(=NCC(=O)[O-])C1=CC=CC=C1N=C(C2=CC=CC=N2)[O-].[Ni+2]
353.991
353.99

Chemical Identifiers

264921-97-3
353.991
53384302
CC(=NCC(=O)[O-])C1=CC=CC=C1N=C(C2=CC=CC=N2)[O-].[Ni+2]
C16H13N3NiO3
SNANRKUUXQKAKV-UHFFFAOYSA-L
nickel(2+);2-[1-[2-[[oxido(pyridin-2-yl)methylidene]amino]phenyl]ethylideneamino]acetate
Safety and Handling

Safety and Handling

TSCA : No

Documents