missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mystaire™ CA Series Cyanoacrylate Fuming Chambers

Safely fume evidence with consistent, reproducible results with the Mystaire CA Series Cyanoacrylate Fuming Chamber—a fully automated cyanoacrylate fingerprinting system.

Manufacturer:  Mystaire™ CA3000

 View more versions of this product

Catalog No. 15-338-306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Extensive Safety Features

  • Patented recirculatory ductless filtration
  • Door automatically locks during fuming
  • Seamless gasket ensures door is sealed
  • Custom filter for cyanoacrylate fumes
  • 500 cycles per cyanoacrylate filter
  • 50 cycles per prefilter
Precision Controls
  • Fully automated system
  • Accurate humidity control to 80%
  • Customizable fuming and filtering intervals
  • Display of options and parameters
  • Programmable internal circulation fan
  • High-temperature hotplate, 160°C
Superior Design and Construction
  • Powder-coated aluminum and steel frame
  • Safety glass door
  • Easy-to-clean interior
  • Sealed internal fluorescent lighting
  • Customizable shelving and interior features

CA-3000 and CA-6000 Models

  • Tabletop ductless units
  • Humidity control, heat source, internal circulation fan
  • Microprocessor control
  • Carbon filtering system captures hazardous fumes
  • All systems ship complete with filters
CA-9000 Model
  • High-volume, stand-alone fingerprinting chamber
  • Can be ductless or ducted to outside
  • Designed to U. S. government standards
  • Ideal for processing large amounts of evidence on a regular basis
  • Accommodates doors, tires, bicycles, tents and other large items

Order Info

For 110V 60Hz or 220V 50Hz.

Specifications

Specifications

25 in.
32 in.
39 in.
50/60Hz
320°F
110/220V
30 in.
99cm
110/220V 50/60Hz
63.5cm
53 in.
36 in.
134.6cm
160°C
81.3cm
76.2cm
32 x 39 x 53 in. (81.3 x 99 x 134.6cm)
Documents