Please login to your online account to display your discounted pricing
Thermo Scientific™ mySPIN™ Mini Centrifuge Series Promotion
Bring increased efficiency and user comfort to your lab with Thermo Scientific™ mySPIN™ 6 and 12 mini centrifuges.
$778.38 - $1,711.75
Specifications
Certifications/Compliance CE, CSA, UL
Display Digital
Electrical Requirements 110-240V 50-60Hz with 4 interchangeable plug adapters
Noise Level 51-53 dBA
Warranty 2 years
View More Specs
Products
Catalog Number Mfr. No. Includes Model Capacity Price Quantity    
75-004-061PMO
thermo scientific™
75004061P
6 x 1.5/2.0mL tube rotor, 16 x 0.2mL tube rotor (singles or 2 x 8 strip), 6 tube adapters 0.2mL, 6 tube adapters 0.5mL, storage case for rotors and adapters, tube rack, universal power adapter 110-240V 50/60 Hz, with 4 interchangeable plug type mySPIN 6 Mini Centrifuge 6 x 2mL Each for $778.38
75-004-081PMO
thermo scientific™
75004081P
12 x 1.5/2.0mL tube rotor, 32 x 0.2mL tube rotor (singles or 4 x 8 strip), 12 tube adapters 0.2mL, 12 tube adapters 0.5mL, universal power adapter 110-240V 50/60 Hz, with 4 interchangeable plug type mySPIN 12 Mini Centrifuge 12 x 2mL Each for $1,711.75
Description & Specifications
Specifications
Certifications/Compliance CE, CSA, UL
Display Digital
Electrical Requirements 110-240V 50-60Hz with 4 interchangeable plug adapters
Noise Level 51-53 dBA
Warranty 2 years
Bring increased efficiency and user comfort to your lab with Thermo Scientific™ mySPIN™ 12 and 6 mini centrifuges. Tool-free rotors and quick-spin options make them fast and easy to use. Benefit from a compact footprint that fits into your workstation and ergnonomic design that stands out. Ideal for microfilter cell separation and HPLC protocols, with the mySPIN 12 centrifuge also supporting PCR, as well as clinical and diagnostic applications.
Benefit from:
  • Compact footprint
  • Easy operation
  • Quiet operation, low vibration
  • Tool-free, quick rotor exchange
  • Quick-spin feature
mySPIN 12 mini centrifuge also features:
  • Imbalance and tilt safety cut-out, as well as safety interlock lid designed to prevent opening while rotor is turning
  • Ability to perform fast spins with higher RCF requirements
  • Back-lit, customizable display that can be set for rpm or RCF
mySPIN 6 mini centrifuge also offers:
  • Safety features: unit will not operate without lid in place
  • Palm-shaped lid for left- or right-handed users
  • Quick tube extraction
Applications:
  • mySPIN 12 centrifuge is ideal for most protocols requiring fast spins (12,500rpm/9,800 x g), and appropriate for PCR, microfilter cell separation, and HPLC protocols
  • mySPIN 6 is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2mL)