Please login to your online account to display your discounted pricing
Thermo Scientific™ mySPIN™ Mini Centrifuge Series
Bring increased efficiency and user comfort to your lab with Thermo Scientific™ mySPIN™ 6 and 12 mini centrifuges.
$820.43 - $1,780.98
Specifications
Certifications/Compliance CE, CSA, UL
Electrical Requirements 110-240V 50-60Hz with 4 interchangeable plug adapters
Noise Level 51-53 dBA
Refrigerated No
Warranty 2 years
View More Specs
Products
Catalog Number Mfr. No. Description Includes Capacity Price Quantity    
75-004-081
thermo scientific™
75004081
mySPIN 12 Mini Centrifuge 12 x 1.5/2.0mL rotor, 32 x 0.2mL rotor (singles or 4 x 8 strip), 0.2mL adapters (set of 12), 0.5mL adapters (set of 12), Universal power adapter 110-240V 50/60 Hz with 4 interchangeable plug types 12 x 2mL Each for $1,780.98
75-004-061
thermo scientific™
75004061
mySPIN 6 Mini Centrifuge 6 x 1.5/2.0mL rotor, 16 x 0.2mL rotor (singles or 2 x 8 strip), 0.2mL adapters (set of 6), 0.5mL adapters (set of 6), Tube rack, Universal power adapter 110-240V 50/60 Hz with 4 interchangeable plug types 6 x 2mL Each for $820.43
Description & Specifications
Specifications
Certifications/Compliance CE, CSA, UL
Electrical Requirements 110-240V 50-60Hz with 4 interchangeable plug adapters
Noise Level 51-53 dBA
Refrigerated No
Warranty 2 years
Bring increased efficiency and user comfort to your lab with Thermo Scientific™ mySPIN™ 12 and 6 mini centrifuges. Tool-free rotors and quick-spin options make them fast and easy to use. Benefit from a compact footprint that fits into your workstation and ergnonomic design that stands out. Ideal for microfilter cell separation and HPLC protocols, with the mySPIN 12 centrifuge also supporting PCR, as well as clinical and diagnostic applications.
Benefit from:
  • Compact footprint
  • Easy operation
  • Quiet operation, low vibration
  • Tool-free, quick rotor exchange
  • Quick-spin feature
mySPIN 12 mini centrifuge also features:
  • Imbalance and tilt safety cut-out, as well as safety interlock lid designed to prevent opening while rotor is turning
  • Ability to perform fast spins with higher RCF requirements
  • Back-lit, customizable display that can be set for rpm or RCF
mySPIN 6 mini centrifuge also offers:
  • Safety features: unit will not operate without lid in place
  • Palm-shaped lid for left- or right-handed users
  • Quick tube extraction
Applications:
  • mySPIN 12 centrifuge is ideal for most protocols requiring fast spins (12,500rpm/9,800 x g), and appropriate for PCR, microfilter cell separation, and HPLC protocols
  • mySPIN 6 is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2mL)
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
2 years