Please login to your online account to display your discounted pricing

(-)-Myrtenal, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (-)-Myrtenal
Name Note 98%
CAS 18486-69-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H14O
Formula Weight 150.22
MDL Number MFCD00074768
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC203620500

  SDS
thermo fisher scientific
203620500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC203620100

  SDS
thermo fisher scientific
203620100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 150.22
Quantity 50g
Color Clear colorless to yellow
EINECS Number 209-274-8
TSCA TSCA
Density 0.98g/mL
Specific Rotation Condition −15° (22°C neat)
RTECS Number DT5180000
Beilstein 07, 161
CAS Max % 100.0
Chemical Name or Material (-)-Myrtenal
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C10H14O
MDL Number MFCD00074768
CAS 18486-69-6
Packaging Glass bottle
CAS Min % 97.5
Refractive Index 1.5029 to 1.5049 (20°C, 589nm)
Specific Rotation -14° to -16° (22°C, 589nm) (neat)