missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 629-63-0 |
---|---|
Molecular Formula | C14H27N |
Molecular Weight (g/mol) | 209.377 |
MDL Number | MFCD00019848 |
InChI Key | MLRCLPRHEOPXLL-UHFFFAOYSA-N |
Synonym | myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3 |
PubChem CID | 64678 |
IUPAC Name | tetradecanenitrile |
SMILES | CCCCCCCCCCCCCC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M074025ML |
TCI America
M074025ML |
25mL |
Each for $56.53
|
|
M0740500ML |
TCI America
M0740500ML |
500mL |
Each for $559.17
|
|
Chemical Identifiers
629-63-0 | |
209.377 | |
MLRCLPRHEOPXLL-UHFFFAOYSA-N | |
64678 | |
CCCCCCCCCCCCCC#N |
C14H27N | |
MFCD00019848 | |
myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3 | |
tetradecanenitrile |
Specifications
227°C | |
25mL | |
C14H27N | |
myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3 | |
CCCCCCCCCCCCCC#N | |
209.377 | |
209.38 | |
≥96.0% (GC) |
Yellow | |
629-63-0 | |
MFCD00019848 | |
MLRCLPRHEOPXLL-UHFFFAOYSA-N | |
tetradecanenitrile | |
64678 | |
Clear Liquid at 20°C | |
Myristonitrile |
Safety and Handling
TSCA : Yes