missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Myristic Anhydride, TCI America™

$230.70 - $1,210.03

Chemical Identifiers

CAS 626-29-9
Molecular Formula C28H54O3
Molecular Weight (g/mol) 438.737
MDL Number MFCD00056151
InChI Key RCRYHUPTBJZEQS-UHFFFAOYSA-N
PubChem CID 69377
IUPAC Name tetradecanoyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M048425G View Documents TCI America
M048425G
25g Each for $230.70
 
M0484250G View Documents TCI America
M0484250G
250g Each for $1,210.03
 
Specifications

Chemical Identifiers

626-29-9
438.737
RCRYHUPTBJZEQS-UHFFFAOYSA-N
tetradecanoyl tetradecanoate
C28H54O3
MFCD00056151
69377
CCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCC

Specifications

White
25g
C28H54O3
RCRYHUPTBJZEQS-UHFFFAOYSA-N
tetradecanoyl tetradecanoate
69377
Crystal-Powder at 20°C
Myristic Anhydride
54°C
626-29-9
MFCD00056151
CCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCC
438.737
438.74
≥95.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes