missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Myricitrin, TCI America™

$84.03 - $282.64

Chemical Identifiers

CAS 17912-87-7
Molecular Formula C21H20O12
Molecular Weight (g/mol) 464.379
MDL Number MFCD00016930
InChI Key DCYOADKBABEMIQ-OWMUPTOHSA-N
Synonym 3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-Rhamnoside
PubChem CID 5281673
ChEBI CHEBI:70082
IUPAC Name 5,7-dihydroxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
SMILES CC1C(C(C(C(O1)OC2=C(OC3=CC(=CC(=C3C2=O)O)O)C4=CC(=C(C(=C4)O)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M236110MG View Documents TCI America
M236110MG
10mg Each for $84.03
 
M236150MG View Documents TCI America
M236150MG
50mg Each for $282.64
 
Specifications

Chemical Identifiers

17912-87-7
464.379
DCYOADKBABEMIQ-OWMUPTOHSA-N
5281673
5,7-dihydroxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
C21H20O12
MFCD00016930
3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-Rhamnoside
CHEBI:70082
CC1C(C(C(C(O1)OC2=C(OC3=CC(=CC(=C3C2=O)O)O)C4=CC(=C(C(=C4)O)O)O)O)O)O

Specifications

Undesignated
10mg
C21H20O12
3,3′C,4′C,5,5′C,7-Hexahydroxyflavone 3-O-Rhamnoside
CC1C(C(C(C(O1)OC2=C(OC3=CC(=CC(=C3C2=O)O)O)C4=CC(=C(C(=C4)O)O)O)O)O)O
464.379
CHEBI:70082
Crystal-Powder at 20°C
Myricitrin
197°C
17912-87-7
MFCD00016930
DCYOADKBABEMIQ-OWMUPTOHSA-N
5,7-dihydroxy-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
5281673
464.38
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator