missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Myricetin, 98%

$73.57 - $262.16

Chemical Identifiers

CAS 529-44-2
Molecular Formula C15H10O8
Molecular Weight (g/mol) 318.237
MDL Number MFCD00006827
InChI Key IKMDFBPHZNJCSN-UHFFFAOYSA-N
Synonym myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
PubChem CID 5281672
ChEBI CHEBI:18152
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
SMILES C1=C(C=C(C(=C1O)O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60450MA View Documents Alfa Aesar™
J60450MA
10mg Each for $73.57
 
AAJ60450MB View Documents Alfa Aesar™
J60450MB
25mg Each for $137.88
 
AAJ60450MF View Documents Alfa Aesar™
J60450MF
50mg Each for $262.16
 
Description

Description

Applications
Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide, dimethyl formamide and ethanol. Insoluble in water.  

Notes
Store in a cool place. Incompatible with strong oxidizing agents.    
Specifications

Chemical Identifiers

529-44-2
318.237
IKMDFBPHZNJCSN-UHFFFAOYSA-N
5281672
3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
C15H10O8
MFCD00006827
myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
CHEBI:18152
C1=C(C=C(C(=C1O)O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O

Specifications

Brown-Yellow
10mg
C15H10O8
332331
myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
IKMDFBPHZNJCSN-UHFFFAOYSA-N
3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
5281672
318.24
98%
>300°C
529-44-2
MFCD00006827
14,6332
Soluble in DMSO or DMF at approximately 10mg/ml; Also soluble in ethanol at 1mg/ml
C1=C(C=C(C(=C1O)O)O)C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O
318.237
CHEBI:18152
Crystalline Solid
Myricetin
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C