missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Myrcene (stabilized with BHT), TCI America™

Manufacturer:  TCI America M0235500ML

 View more versions of this product

Catalog No. M0235500ML



Specifications

Specifications

Myrcene (stabilized with BHT)
Yellow
123-35-3
MFCD00008908
myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural
CC(=CCCC(=C)C=C)C
136.238
CHEBI:17221
Clear Liquid at 20°C
167°C
500mL
C10H16
3295
UAHWPYUMFXYFJY-UHFFFAOYSA-N
7-methyl-3-methylideneocta-1,6-diene
31253
136.24
≥75.0% (GC)

Chemical Identifiers

123-35-3
136.238
UAHWPYUMFXYFJY-UHFFFAOYSA-N
31253
7-methyl-3-methylideneocta-1,6-diene
C10H16
MFCD00008908
myrcene, beta-myrcene, 7-methyl-3-methyleneocta-1,6-diene, 7-methyl-3-methylene-1,6-octadiene, 1,6-octadiene, 7-methyl-3-methylene, beta-geraniolene, .beta.-myrcene, 2-methyl-6-methylene-2,7-octadiene, 3-methylene-7-methyl-1,6-octadiene, myrcene natural
CHEBI:17221
CC(=CCCC(=C)C=C)C
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2358

RTECSNumber : RG5365000

TSCA : Yes

Recommended Storage : Refrigerator

Documents