Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody.

$236.94 - $1,192.98

Specifications

Antigen Myosin, Smooth Muscle Heavy Chain Ab-3
Clone SM-M10
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1348P

 
thermo scientific™ lab vision™
MS1348P
Purified Ab with BSA and Azide 1mL Each for $1,110.06

MS1348P0

 
thermo scientific™ lab vision™
MS1348P0
Purified Ab with BSA and Azide 100µL Each for $236.94

MS1348P1

 
thermo scientific™ lab vision™
MS1348P1
Purified Ab with BSA and Azide 500µL Each for $728.65

MS1348P1ABX

 
thermo scientific™ lab vision™
MS1348P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $815.86

MS1348PABX

 
thermo scientific™ lab vision™
MS1348PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,192.98
Description & Specifications

Specifications

Antigen Myosin, Smooth Muscle Heavy Chain Ab-3
Clone SM-M10
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Bovine
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Type Primary
Immunogen Crude human uterus extract
Isotype IgG2a kappa

Provides accurate, reproducible results with the Thermo Scientific™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody.

SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.

Host Species: Mouse

Clone: SM-M10

Isotype: IgG1/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 205kDa (MHC-1) and 200kDa (MHC-2)

Positive Control: Uterus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence
  • Immunoprecipitation (Not verified)
Note:

USA: RUO; Int'l: RUO