Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody


Provides accurate, reproducible results with the Thermo Scientific™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1348PABX

 View more versions of this product

Catalog No. MS1348PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Myosin, Smooth Muscle Heavy Chain Ab-3
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SM-M10
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Crude human uterus extract
Isotype IgG2a kappa
Quantity 200μL
Regulatory Status RUO
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Bovine
Target Species Human

Provides accurate, reproducible results with the Thermo Scientific™ Myosin, Smooth Muscle Heavy Chain Ab-3, Mouse Monoclonal Antibody.

SM-MHC is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmently regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2.

Host Species: Mouse

Clone: SM-M10

Isotype: IgG1/κ

Species Reactivity: Human and Cow. Others not known.

Epitope: Not determined

Immunogen: Crude human uterus extract

Molecular Weight: 205kDa (MHC-1) and 200kDa (MHC-2)

Positive Control: Uterus

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence
  • Immunoprecipitation (Not verified)
Note:

USA: RUO; Int'l: RUO