Please login to your online account to display your discounted pricing

Myosin Mouse, Alexa Fluor 488, Clone: SMMS-1, eBioscience™

Mouse Monoclonal Antibody

$175.45 - $413.25

Specifications

Antigen Myosin
Clone SMMS-1
Host Species Murine
Gene Alias smooth muscle myosin heavy chains, SM1 and SM2 isoforms
Species Reactivity Bovine, Canine, cat, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124606

 
life technologies
LS53640080
25μg Each for $175.45

501124607

 
life technologies
LS53640082
100μg Each for $413.25
Description & Specifications

Specifications

Antigen Myosin
Clone SMMS-1
Host Species Murine
Gene Alias smooth muscle myosin heavy chains, SM1 and SM2 isoforms
Species Reactivity Bovine, Canine, cat, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This SMMS-1 monoclonal antibody reacts with both isoforms of the human smooth muscle myosin heavy chain (SM1 and SM2) structural proteins, which are components of the contractile apparatus in smooth muscle cells. Smooth muscle myosin heavy chains are expressed by myoepithelial cells and also serve as the terminal differentiation marker for smooth muscle cells. In normal breast tissue, myoepithelial cells compose the outer layer of the lumen lining. In benign and in situ breast lesions, the intact myoepithelial layer stains with SMMS-1 and the loss of this staining indicates invasive cancer. This SMMS-1 antibody has also been reported to cross-react with bovine, canine, feline, and rat smooth muscle myosin heavy chains.