Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Myoglobin, Rabbit Polyclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ Myoglobin, Rabbit Polyclonal Antibody.

$393.76 - $769.04

Specifications

Antigen Myoglobin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Format Quantity Price Quantity    

RB9068P

 
thermo scientific™ lab vision™
RB9068P
Concentrated 1mL Each for $769.04

RB9068P0

 
thermo scientific™ lab vision™
RB9068P0
Concentrated 100μL Each for $393.76

RB9068P1

 
thermo scientific™ lab vision™
RB9068P1
Concentrated 500μL Each for $495.02

RB9068R7

 
thermo scientific™ lab vision™
RB9068R7
Prediluted 7mL Each for $500.57
Description & Specifications

Specifications

Antigen Myoglobin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Rabbit
Regulatory Status RUO
Research Discipline Muscle Biology
Type Primary
Immunogen Purified human myoglobin
Isotype IgG

Provides accurate, reproducible results with the Thermo Scientific™ Myoglobin, Rabbit Polyclonal Antibody.

Myoglobin is a cytoplasmic, single chain polypeptide of 153 amino acids and contains a single heme group. Myoglobin is found in skeletal and carciac muscle but not in smooth muscle and functions as an oxygen transporting pigment. Upon damage to the muscle cell due to infarction of a coronary artery, neurological trauma, infection of tumor processes, myoglobin escapes to the environment and can be found in plasma using sensitive assays.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human myoglobin

Molecular Weight: 17.8kDa

Positive Control: Skeletal muscle

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO