Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Myogenin Ab-2, Mouse Monoclonal Antibody

Provides accurate, reproducible results with the Thermo Scientific™ Myogenin Ab-2, Mouse Monoclonal Antibody.

$244.37 - $1,204.57

Specifications

Antigen Myogenin Ab-2
Clone F12B
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS1182P

 
thermo scientific™ lab vision™
MS1182P
Purified Ab with BSA and Azide 1mL Each for $1,117.35

MS1182P0

 
thermo scientific™ lab vision™
MS1182P0
Purified Ab with BSA and Azide 100μL Each for $244.37

MS1182P1

 
thermo scientific™ lab vision™
MS1182P1
Purified Ab with BSA and Azide 500μL Each for $709.55

MS1182P1ABX

 
thermo scientific™ lab vision™
MS1182P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $795.55

MS1182PABX

 
thermo scientific™ lab vision™
MS1182PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,204.57
Description & Specifications

Specifications

Antigen Myogenin Ab-2
Clone F12B
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Muscle Biology
Type Primary
Immunogen Recombinant protein containing rat myogenin aa 30-224
Isotype IgG2a kappa

Provides accurate, reproducible results with the Thermo Scientific™ Myogenin Ab-2, Mouse Monoclonal Antibody.

Myogenin is a member of a family of myogenic regulatory genes, which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors, which are essential for muscle development. Transfection of myogenin into multipotential mesodermal cells have been shown to convert the mesodermal cells to myoblasts. Expression of myogenin is restricted to cells of skeletal muscle origin. It is therefore a useful marker for tumors of the muscle lineage, being strongly expressed in alveolar rhabdomyosarcomas.

Host Species: Mouse

Clone: F12B

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 158-178

Immunogen: Recombinant protein containing rat myogenin aa 30-224

Molecular Weight: 34kDa

Positive Control: Rh30 cells; Rhabdomyosarcoma or Wilm's Tumor

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO