Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Myogenin Ab-2, Mouse Monoclonal Antibody


Provides accurate, reproducible results with the Thermo Scientific™ Myogenin Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1182P1ABX

 View more versions of this product

Catalog No. MS1182P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Myogenin Ab-2
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Clone F12B
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Murine
Immunogen Recombinant protein containing rat myogenin aa 30-224
Isotype IgG2a kappa
Quantity 100μL
Regulatory Status RUO
Research Discipline Muscle Biology
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human
Target Species Murine
Target Species Rat

Provides accurate, reproducible results with the Thermo Scientific™ Myogenin Ab-2, Mouse Monoclonal Antibody.

Myogenin is a member of a family of myogenic regulatory genes, which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors, which are essential for muscle development. Transfection of myogenin into multipotential mesodermal cells have been shown to convert the mesodermal cells to myoblasts. Expression of myogenin is restricted to cells of skeletal muscle origin. It is therefore a useful marker for tumors of the muscle lineage, being strongly expressed in alveolar rhabdomyosarcomas.

Host Species: Mouse

Clone: F12B

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 158-178

Immunogen: Recombinant protein containing rat myogenin aa 30-224

Molecular Weight: 34kDa

Positive Control: Rh30 cells; Rhabdomyosarcoma or Wilm's Tumor

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO