missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ MyoD1 Ab-2, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ MyoD1 Ab-2, Mouse Monoclonal Antibody.

$243.41 - $1,059.43

Specifications

Antigen MyoD1 Ab-2
Clone 5.2F
Applications Immunofluorescence,Immunoprecipitation,Immunofluorescence,Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS278P Thermo Scientific™
MS278P
200μg/mL 1mL Each for $943.51
MS278P0 Thermo Scientific
MS278P0
200μg/mL 100μL Each for $243.41
MS278P1 Thermo Scientific™
MS278P1
200μg/mL 500μL Each for $706.88
MS278P1ABX Thermo Scientific™
MS278P1ABX
1mg/mL 100μL Each for $800.04
MS278PABX Thermo Scientific™
MS278PABX
1mg/mL 200μL Each for $1,059.43
Specifications

Description

Deliver accurate, reproducible results with the Thermo Scientific™ MyoD1 Ab-2, Mouse Monoclonal Antibody.

Research Areas: Muscle Markers; MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occassionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm's tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing's sarcomas and alveolar soft part sarcomas.

Host Species: Mouse

Clone: 5.2F

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Chicken. Others not known.

Epitope: aa 3-56

Immunogen: Recombinant mouse MyoD1 protein

Molecular Weight: 45kDa

Positive Control: SW80 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

MyoD1 Ab-2
Immunofluorescence,Immunoprecipitation,Immunofluorescence,Immunoprecipitation
Unlabeled
Murine
Muscle Biology
Recombinant mouse MyoD1 protein
5.2F
Monoclonal
Concentrated
RUO
Primary
IgG2a kappa