Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MyoD1 Ab-1, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ MyoD1 Ab-1, Mouse Monoclonal Antibody.

$248.82 - $1,029.35

Specifications

Antigen MyoD1 Ab-1
Clone 5.8A
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS273P

 
thermo scientific™ lab vision™
MS273P
200μg/mL 1mL Each for $912.20

MS273P0

 
thermo scientific™ lab vision™
MS273P0
200μg/mL 100µL Each for $248.82

MS273P1

 
thermo scientific™ lab vision™
MS273P1
200μg/mL 500µL Each for $681.65

MS273P1ABX

 
thermo scientific™ lab vision™
MS273P1ABX
1mg/mL 100µL Each for $756.94

MS273PABX

 
thermo scientific™ lab vision™
MS273PABX
1mg/mL 200µL Each for $1,029.35
Description & Specifications

Specifications

Antigen MyoD1 Ab-1
Clone 5.8A
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Immunofluorescence
Applications Immunoprecipitation
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Muscle Biology
Type Primary
Immunogen Recombinant mouse MyoD1 protein
Isotype IgG2a kappa

Deliver accurate, reproducible results with the Thermo Scientific™ MyoD1 Ab-1, Mouse Monoclonal Antibody.

MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occassionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm's tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing's sarcomas and alveolar soft part sarcomas.

Host Species: Mouse

Clone: 5.8A

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, Chicken.

Epitope: aa 180-189

Immunogen: Recombinant mouse MyoD1 protein

Molecular Weight: 45kDa

Positive Control: SW80 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO