Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MyoD1 Ab-1, Mouse Monoclonal Antibody


Deliver accurate, reproducible results with the Thermo Scientific™ MyoD1 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS273P1ABX

 View more versions of this product

Catalog No. MS273P1ABX

  • / Each
 

Description & Specifications

Specifications

Quantity 100µL
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunofluorescence
Applications Immunoprecipitation
Regulatory Status RUO
Antigen MyoD1 Ab-1
Immunogen Recombinant mouse MyoD1 protein
Clone 5.8A
Format Concentrated
Host Species Mouse
Isotype IgG2a kappa
Research Discipline Muscle Biology
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Deliver accurate, reproducible results with the Thermo Scientific™ MyoD1 Ab-1, Mouse Monoclonal Antibody.

MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occassionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm's tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing's sarcomas and alveolar soft part sarcomas.

Host Species: Mouse

Clone: 5.8A

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, Chicken.

Epitope: aa 180-189

Immunogen: Recombinant mouse MyoD1 protein

Molecular Weight: 45kDa

Positive Control: SW80 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO