Invitrogen™ MYLK2 (skMLCK) Recombinant Human Protein DFS Item
MYLK2 (skMLCK) is a calcium/calmodulin-dependent serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4025
 View more versions of this product
Catalog No. PV4025
Description
Description
MYLK2 (skMLCK) is expressed exclusively in adult skeletal muscle.
Specifications
Specifications
NP_149109
Frozen
92.1kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
MLCK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MYLK2 (skMLCK)
Baculovirus
Human
GST Tag
Documents