missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Myeloperoxidase (MPO), Rabbit Polyclonal Antibody

Myeloperoxidase (MPO), Rabbit Polyclonal Ab is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

$211.84 - $828.98

Specifications

Antigen Myeloperoxidase (MPO)
Clone Polyclonal
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RB373A Thermo Scientific™ Lab Vision™
RB373A
1mL Each for $828.98
RB373A0 Thermo Scientific™ Lab Vision™
RB373A0
100μL Each for $211.84
RB373A1 Thermo Scientific™ Lab Vision™
RB373A1
500μL Each for $599.86
RB373R7 Thermo Scientific™ Lab Vision™
RB373R7
7mL Each for $344.53
Specifications

Description

Choose one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available with the Thermo Scientific™ Myeloperoxidase (MPO), Rabbit Polyclonal Antibody.

Myeloperoxidase is an important enzyme used by granulocytes during phagocytic lysis of foreign particles engulfed. In normal tissues and in a variety of myeloproliferative disorders myeloid cells of both neutrophilic and eosinophilic types, at all stages of maturation, exhibit strong cytoplasmic reactivity for MPO. Erythroid precursors, megakaryocytes, lymphoid cells, mast cells, and plasma cells are nonreactive. MPO is not observed in the neoplastic cells of a wide variety of epithelial tumors and sarcomas. MPO is useful in differentiating between myeloid and lymphoid leukemias.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Purified human granulocytic MPO

Positive Control: Tonsil or spleen

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Description & Specifications

Specifications

Myeloperoxidase (MPO)
Immunohistochemistry (Paraffin)
Unlabeled
IVD
Primary
Polyclonal
Polyclonal
Rabbit
Enzyme
Purified human granulocytic MPO