missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Myeloperoxidase (MPO) Mouse anti-Human, PE, Clone: MPO455-8E6, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $381.48

Specifications

Antigen Myeloperoxidase (MPO)
Clone MPO455-8E6
Host Species Murine
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245284 Life Technologies
LS12129941
25 Tests Each for $157.08
50245285 Life Technologies
LS12129942
100 Tests Each for $381.48
Specifications

Description

The monoclonal antibody MPO455-8E6 recognizes myeloperoxidase (MPO), a protein within the azurophilic granules of myeloid cells. MPO is a multimeric protein comprised of two 55kDa subunits and two 15kDa subunits. The larger subunits associate with a heme protein resulting in a greenish color. As an enzyme, MPO breaks down hydrogen peroxide and oxidizes tyrosine. The products of this reaction, hypochlorous acid and tyrosyl radical, cause the cytotoxic and killing effects characteristic of neutrophils. Myeloperoxidase is important in the diagnosis of some cancers and increases in serum levels have been shown to correlate with cardiac events.
Description & Specifications

Specifications

Myeloperoxidase (MPO)
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MPO455-8E6
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test