Please login to your online account to display your discounted pricing

Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts


Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.

Manufacturer: thermo scientific™  PIEW84857

 View more versions of this product

Catalog No. PIEW84857

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) myECL Imager
Description Extended Warranty
Quantity 1 warranty
Sufficient For Covers all parts (non-consumable) and labor; includes shipping and handling
Item Description Extended Warranty
Formulation One year extended warranty

Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.

Replacement parts for the myECL Imager and accessories to expand upon its capabilities.