Please login to your online account to display your discounted pricing

Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts


Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.

Manufacturer: thermo scientific™  62267

 View more versions of this product

Catalog No. PI62267

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) myECL™ Imager
For Use With (Application) myECL Imager
Description Imaging Reference Target
Formulation Plastic imaging target with printed design; 7 x 6.5in.
Quantity 1 sheet
Sufficient For Replacement of myECL Imager accessory; used for focusing in all imaging modes
Item Description Imaging Reference Target

Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.

Replacement parts for the myECL Imager and accessories to expand upon its capabilities.